Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC19A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025243.3 | NP_079519.1 | 3 | 401 | ||
XM_005246874.3 | XP_005246931.1 | 4 | 564 | |||
XM_011511931.2 | XP_011510233.1 | 3 | 656 | |||
XM_011511932.1 | XP_011510234.1 | 3 | 478 | |||
XM_011511933.1 | XP_011510235.1 | 3 | 744 | |||
XM_017005030.1 | XP_016860519.1 | 4 | 746 | |||
XM_017005031.1 | XP_016860520.1 | 4 | 725 | |||
XM_017005032.1 | XP_016860521.1 | 4 | 606 | |||
XM_017005033.1 | XP_016860522.1 | 4 | 920 | |||
XM_017005034.1 | XP_016860523.1 | 4 | 850 | |||
GenBank mRNA | AF271633.1 | - | 3 | 355 | ||
AF283317.1 | - | 3 | 382 | |||
AK301490.1 | - | 4 | 567 | |||
AK312464.1 | - | 3 | 403 | |||
BC032014.1 | - | 3 | 356 | 5177540 |