Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136538.1 | NP_001130010.1 | 14, 15 | 2416 | ||
NM_025247.5 | NP_079523.3 | 13, 14 | 2323 | |||
GenBank mRNA | AK092356.1 | - | 13, 14 | 2308 | ||
AK097425.1 | - | 7, 8 | 1273 | |||
AL832043.1 | - | 14, 15 | 2394 | |||
AY323912.1 | - | 13, 14 | 2279 | |||
BC003698.1 | - | 9, 10 | 1480 | |||
BC126358.1 | - | 13, 14 | 2237 | 8992074 |