Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270367.1 | NP_001257296.1 | 4 | 522 | ||
NM_001270368.1 | NP_001257297.1 | 4 | 535 | |||
NM_001270369.1 | NP_001257298.1 | 5 | 574 | |||
NM_001270370.1 | NP_001257299.1 | 6 | 581 | |||
NM_001270371.1 | NP_001257300.1 | 5 | 596 | |||
NM_001270372.1 | NP_001257301.1 | 4 | 514 | |||
NM_001270373.1 | NP_001257302.1 | 5 | 553 | |||
NM_001270374.1 | NP_001257303.1 | 5 | 903 | |||
NM_025125.3 | NP_079401.2 | 4 | 501 | |||
NR_072984.1 | 5 | 518 | ||||
NR_072985.1 | 5 | 525 | ||||
NR_072986.1 | 4 | 423 | ||||
NR_072987.1 | 5 | 475 | ||||
XM_006717978.2 | XP_006718041.1 | 5 | 853 | |||
GenBank mRNA | AK023325.1 | - | 4 | 477 | ||
AK222536.1 | - | 4 | 482 | |||
AK300490.1 | - | 4 | 532 | |||
AK309425.1 | - | 5 | 800 | |||
AK309980.1 | - | 4 | 509 | |||
AK310160.1 | - | 5 | 483 | |||
AK311563.1 | - | 5 | 491 | |||
AL832397.1 | - | 4 | 398 | |||
AL832483.1 | - | 4 | 509 | |||
BC022252.1 | - | 4 | 477 | 4668550 | ||
BP204746.1 | - | 6 | 550 | |||
BP293793.1 | - | 5 | 528 | |||
BX537803.1 | - | 5 | 483 | |||
CX756014.1 | - | 6 | 537 | |||
DA573956.1 | - | 5 | 534 | |||
DA930892.1 | - | 4 | 490 |