Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270367.1 | NP_001257296.1 | 4 | 1293 | ||
NM_001270368.1 | NP_001257297.1 | 4 | 1306 | |||
NM_001270369.1 | NP_001257298.1 | 5 | 1345 | |||
NM_001270370.1 | NP_001257299.1 | 6 | 1352 | |||
NM_001270371.1 | NP_001257300.1 | 5 | 1367 | |||
NM_001270372.1 | NP_001257301.1 | 4 | 1285 | |||
NM_001270373.1 | NP_001257302.1 | 5 | 1324 | |||
NM_001270374.1 | NP_001257303.1 | 5 | 1674 | |||
NM_025125.3 | NP_079401.2 | 4 | 1272 | |||
NR_072984.1 | 5 | 1289 | ||||
NR_072985.1 | 5 | 1296 | ||||
NR_072986.1 | 4 | 1194 | ||||
NR_072987.1 | 5 | 1246 | ||||
XM_006717978.2 | XP_006718041.1 | 5 | 1624 | |||
GenBank mRNA | AK023325.1 | - | 4 | 1248 | ||
AK222536.1 | - | 4 | 1253 | |||
AK300490.1 | - | 4 | 1303 | |||
AL832397.1 | - | 4 | 1169 | |||
AL832483.1 | - | 4 | 1280 | |||
BC022252.1 | - | 4 | 1248 | 4668550 | ||
BX537803.1 | - | 5 | 1254 |
Gene Symbol: RAB6C-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036537.1 | 6 | 1645 | |||
GenBank mRNA | AK123452.1 | - | 6 | 1582 | ||
DQ786271.1 | - | 6 | 1651 |