Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REEP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316964.1 | NP_001303893.1 | 7 | 1306 | ||
NM_001316965.1 | NP_001303894.1 | 7 | 1325 | |||
NM_025232.3 | NP_079508.2 | 8 | 1461 | |||
GenBank mRNA | AF218014.1 | - | 8 | 1239 | ||
AK025899.1 | - | 8 | 1391 | |||
AK025930.1 | - | 8 | 1094 | |||
AK172846.1 | - | 8 | 1396 | |||
BC013048.1 | - | 8 | 1445 | 3863838 | ||
BC050622.1 | - | 7 | 1227 | 6504298 |