Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBXN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171091.1 | NP_001164562.1 | 6, 7 | 609 | ||
NM_025241.2 | NP_079517.1 | 6, 7 | 711 | |||
XM_017027325.1 | XP_016882814.1 | 7, 8 | 1142 | |||
GenBank mRNA | AF272893.1 | - | 6, 7 | 710 | ||
AF272894.1 | - | 6, 7 | 600 | |||
AL833695.1 | - | 1, 2 | 566 | |||
BC007414.2 | - | 2, 3 | 171 | |||
BC008288.1 | - | 6, 7 | 609 | 3659797 |