Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030636.2 | NP_085139.2 | 2 | 1173 | ||
XM_017012657.1 | XP_016868146.1 | 2 | 1251 | |||
XM_017012658.1 | XP_016868147.1 | 2 | 1251 | |||
GenBank mRNA | AB051493.1 | - | 2 | 914 | ||
AK027386.1 | - | 2 | 1145 | |||
BC065518.1 | - | 3 | 1073 | 5755727 |