Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258392.2 | NP_001245321.1 | 3 | 687 | ||
NM_001258394.2 | NP_001245323.1 | 4 | 794 | |||
NM_030813.5 | NP_110440.1 | 3 | 687 | |||
XM_011545289.2 | XP_011543591.1 | 3 | 560 | |||
GenBank mRNA | AK023214.1 | - | 3 | 537 | ||
AK302069.1 | - | 4 | 492 | |||
AL136909.1 | - | 3 | 586 | |||
BC006404.2 | - | 3 | 527 | 4304523 | ||
BU848809.1 | - | 3 | 533 |