Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030819.3 | NP_110446.3 | 2, 3 | 555 | ||
XM_017023738.1 | XP_016879227.1 | 2, 3 | 505 | |||
GenBank mRNA | AK074382.1 | - | 3, 4 | 508 | ||
AK225997.1 | - | 2, 3 | 525 | |||
BC002943.1 | - | 1, 2 | 182 | |||
BC107585.1 | - | 2, 3 | 505 | 6463662 | ||
BI826603.1 | - | 2, 3 | 608 |