Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCSTAMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030788.3 | NP_110415.1 | 3 | 1123 | ||
XM_011517321.1 | XP_011515623.1 | 4 | 1492 | |||
XM_017013878.1 | XP_016869367.1 | 5 | 1591 | |||
GenBank mRNA | AF277290.1 | - | 3 | 1101 | ||
AF305068.1 | - | 3 | 1095 | |||
AK095417.1 | - | 3 | 1093 | |||
BC069349.1 | - | 3 | 1093 | 7262322 | ||
BC112018.1 | - | 2 | 1067 | 8327486 | ||
BC112020.1 | - | 2 | 1067 | 8327488 |