Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136043.2 | NP_001129515.1 | 5 | 666 | ||
NM_001207058.1 | NP_001193987.1 | 5 | 645 | |||
NM_001207059.1 | NP_001193988.1 | 4 | 525 | |||
NR_045104.1 | 5 | 809 | ||||
NR_045105.2 | 5 | 1063 | ||||
NR_045106.2 | 5 | 970 | ||||
NR_045901.1 | 4 | 701 | ||||
XM_006720708.3 | XP_006720771.1 | 5 | 745 | |||
XM_006720709.3 | XP_006720772.1 | 5 | 810 | |||
XM_006720711.2 | XP_006720774.1 | 4 | 669 | |||
XM_011522101.2 | XP_011520403.1 | 5 | 491 | |||
XM_017022644.1 | XP_016878133.1 | 5 | 890 | |||
GenBank mRNA | AK027475.1 | - | 5 | 651 | ||
AK293207.1 | - | 4 | 478 | |||
AK296134.1 | - | 5 | 1036 | |||
AK301033.1 | - | 5 | 508 | |||
AK301222.1 | - | 4 | 654 | |||
AK301742.1 | - | 5 | 487 | |||
AL136662.1 | - | 5 | 891 | |||
BC007991.2 | - | 5 | 636 | 3140504 | ||
DC298728.1 | - | 4 | 478 |