Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: EIF2AK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135651.2 | NP_001129123.1 | 3 | 394 | ||
NM_001135652.2 | NP_001129124.1 | 1 | 87 | |||
NM_002759.3 | NP_002750.1 | 3 | 628 | |||
XM_011532987.2 | XP_011531289.1 | 3 | 328 | |||
XM_017004503.1 | XP_016859992.1 | 3 | 324 | |||
GenBank mRNA | AK290655.1 | - | 3 | 314 | ||
AK313818.1 | - | 2 | 151 | |||
AY302136.1 | - | 1 | 71 | |||
BC007769.2 | - | 2 | 170 | |||
BC022314.1 | - | 3 | 282 | |||
BC040851.1 | - | 3 | 394 | |||
BC057805.1 | - | 3 | 299 | |||
BC093676.1 | - | 1 | 99 | 7939521 | ||
BC101475.1 | - | 1 | 99 | 8068981 | ||
BC143228.1 | - | 1 | 98 | |||
BC143230.1 | - | 1 | 99 | 9051731 | ||
M35663.1 | - | 2 | 257 | |||
M85294.1 | - | 1 | 101 |