Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030819.3 | NP_110446.3 | 3 | 1826 | ||
NR_027398.1 | 2 | 1480 | ||||
XM_006721288.3 | XP_006721351.1 | 3 | 1569 | |||
XM_017023738.1 | XP_016879227.1 | 3 | 1776 | |||
GenBank mRNA | AK074382.1 | - | 4 | 1779 | ||
AK091606.1 | - | 1 | 2703 | |||
AK225997.1 | - | 3 | 1796 | |||
AL832904.1 | - | 1 | 1265 | |||
AY358169.1 | - | 2 | 1536 | |||
BC000757.2 | - | 2 | 1433 | 2820364 | ||
BC002943.1 | - | 2 | 1453 | |||
BC107585.1 | - | 3 | 1776 | 6463662 |