Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NIPA2 SIRNA Assays ›
Gene Symbol:
NIPA2
Gene Name:
non imprinted in Prader-Willi/Angelman syndrome 2
Gene Aliases:
-
Chromosome Location:
Chr.15: 22838641 - 22868384 on Build GRCh38
UniGene ID:
Hs.591003
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
32921
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NIPA2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001008860.2
NP_001008860.1
3
637
NM_001008892.2
NP_001008892.1
2
501
NM_001008894.2
NP_001008894.1
2
501
NM_001184888.1
NP_001171817.1
3
649
NM_001184889.1
NP_001171818.1
6
984
NM_030922.6
NP_112184.4
4
759
XM_005272546.3
XP_005272603.1
4
992
XM_005272547.4
XP_005272604.1
4
776
XM_005272548.3
XP_005272605.1
3
606
XM_005272550.3
XP_005272607.1
3
444
XM_005272552.3
XP_005272609.1
2
706
XM_005272553.4
XP_005272610.1
2
510
XM_006720364.2
XP_006720427.1
5
886
XM_006720365.2
XP_006720428.1
4
731
XM_006720366.3
XP_006720429.1
3
597
XM_006720367.1
XP_006720430.1
4
992
XM_011543877.2
XP_011542179.1
5
1119
XM_011543878.2
XP_011542180.1
5
1286
XM_011543879.2
XP_011542181.1
5
857
XM_011543880.2
XP_011542182.1
3
870
XM_017022645.1
XP_016878134.1
5
1180
XM_017022646.1
XP_016878135.1
5
947
XM_017022647.1
XP_016878136.1
5
895
XM_017022648.1
XP_016878137.1
5
956
XM_017022649.1
XP_016878138.1
5
1090
XM_017022650.1
XP_016878139.1
5
1053
XM_017022651.1
XP_016878140.1
6
1474
XM_017022652.1
XP_016878141.1
5
866
XM_017022653.1
XP_016878142.1
6
1180
XM_017022654.1
XP_016878143.1
4
704
XM_017022655.1
XP_016878144.1
4
759
XM_017022656.1
XP_016878145.1
4
769
XM_017022657.1
XP_016878146.1
5
886
XM_017022658.1
XP_016878147.1
3
607
XM_017022659.1
XP_016878148.1
5
857
XM_017022660.1
XP_016878149.1
3
870
XM_017022661.1
XP_016878150.1
3
637
XM_017022662.1
XP_016878151.1
2
706
XM_017022663.1
XP_016878152.1
2
511
GenBank mRNA
AK096305.1
-
3
618
AK300843.1
-
2
444
AK313584.1
-
3
633
AY732242.1
-
6
905
BC011775.2
-
2
413
3640970
BM544162.1
-
4
735
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
magnesium ion transport
magnesium ion transmembrane transport
magnesium ion transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0