Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITM2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012514.2 | NP_001012532.1 | 2 | 346 | ||
NM_001012516.2 | NP_001012534.1 | 3 | 432 | |||
NM_001287240.1 | NP_001274169.1 | 3 | 423 | |||
NM_001287241.1 | NP_001274170.1 | 4 | 619 | |||
NM_030926.5 | NP_112188.1 | 3 | 487 | |||
GenBank mRNA | AB003629.1 | - | 3 | 356 | ||
AF271781.1 | - | 3 | 472 | |||
AF272043.1 | - | 3 | 469 | |||
AK056321.1 | - | 2 | 293 | |||
AK075361.1 | - | 3 | 452 | |||
AK090975.1 | - | 3 | 398 | |||
AK222697.1 | - | 3 | 434 | |||
AK295503.1 | - | 3 | 400 | |||
AK303199.1 | - | 3 | 452 | |||
AK307702.1 | - | 2 | 255 | |||
AL136603.1 | - | 3 | 428 | |||
AL713651.1 | - | 3 | 411 | |||
AY049777.1 | - | 3 | 373 | |||
BC002424.1 | - | 3 | 414 | 3162430 | ||
BC025742.1 | - | 3 | 402 | 5224043 | ||
BC050668.1 | - | 2 | 281 | 5759920 | ||
BC098563.1 | - | 3 | 400 | 30558635 | ||
BM842614.1 | - | 3 | 476 | |||
BX444331.2 | - | 3 | 422 | |||
CR457208.1 | - | 3 | 356 | |||
DA138409.1 | - | 4 | 456 | |||
DA182471.1 | - | 3 | 489 |