Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ABHD17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130111.1 | NP_001123583.1 | 3 | 893 | ||
NM_031213.3 | NP_112490.3 | 4 | 1046 | |||
XM_011528338.1 | XP_011526640.1 | 3 | 3918 | |||
XM_017027341.1 | XP_016882830.1 | 2 | 3765 | |||
XM_017027342.1 | XP_016882831.1 | 2 | 3765 | |||
GenBank mRNA | AK024419.1 | - | 2 | 1651 | ||
AK074043.1 | - | 2 | 2721 | |||
AK074548.1 | - | 3 | 875 | |||
AK090438.1 | - | 2 | 2524 | |||
AK289533.1 | - | 3 | 863 | |||
BC000158.2 | - | 2 | 631 | 2985707 | ||
BC009256.2 | - | 3 | 843 | 3954724 | ||
BC011667.2 | - | 3 | 751 | 4122276 | ||
BC020512.1 | - | 4 | 960 | 3850505 | ||
BC033749.1 | - | 3 | 850 | 5197958 | ||
BC035961.1 | - | 3 | 869 | 4805422 | ||
BC071644.1 | - | 3 | 829 | 5242996 | ||
BC071876.1 | - | 3 | 816 | 6672380 | ||
BC094816.1 | - | 3 | 814 | 6266705 |