Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADPGK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031284.4 | NP_112574.3 | 5 | 915 | ||
NR_023318.1 | 5 | 1128 | ||||
NR_023319.1 | 3 | 626 | ||||
XM_006720712.3 | XP_006720775.1 | 5 | 915 | |||
XM_006720713.3 | XP_006720776.1 | 5 | 1076 | |||
XM_011522104.2 | XP_011520406.1 | 5 | 915 | |||
XM_017022664.1 | XP_016878153.1 | 1 | 856 | |||
GenBank mRNA | AF204270.1 | - | 4 | 504 | ||
AK055526.1 | - | 3 | 617 | |||
AK075560.1 | - | 5 | 850 | |||
AK293282.1 | - | 5 | 607 | |||
AK294391.1 | - | 4 | 652 | |||
AL832254.1 | - | 1 | 527 | |||
BC006112.1 | - | 5 | 857 | 3347312 | ||
BC044589.1 | - | 5 | 1074 |