Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITM2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012514.2 | NP_001012532.1 | 5 | 915 | ||
NM_001012516.2 | NP_001012534.1 | 5 | 890 | |||
NM_001287240.1 | NP_001274169.1 | 6 | 992 | |||
NM_001287241.1 | NP_001274170.1 | 7 | 1188 | |||
NM_030926.5 | NP_112188.1 | 6 | 1056 | |||
GenBank mRNA | AF271781.1 | - | 6 | 1041 | ||
AF272043.1 | - | 6 | 1038 | |||
AK056321.1 | - | 5 | 862 | |||
AK075361.1 | - | 6 | 1021 | |||
AK090975.1 | - | 6 | 967 | |||
AK126739.1 | - | 1 | 2136 | |||
AK222697.1 | - | 6 | 1003 | |||
AK295503.1 | - | 6 | 822 | |||
AK303199.1 | - | 6 | 1010 | |||
AK307702.1 | - | 5 | 824 | |||
AL136603.1 | - | 6 | 997 | |||
AL713651.1 | - | 6 | 980 | |||
BC002424.1 | - | 6 | 983 | 3162430 | ||
BC025742.1 | - | 5 | 860 | 5224043 | ||
BC050668.1 | - | 5 | 850 | 5759920 | ||
BC098563.1 | - | 6 | 969 | 30558635 | ||
CR749389.1 | - | 1 | 2094 |