Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NIPA2 SIRNA Assays ›
Gene Symbol:
NIPA2
Gene Name:
non imprinted in Prader-Willi/Angelman syndrome 2
Gene Aliases:
-
Chromosome Location:
Chr.15: 22838641 - 22868384 on Build GRCh38
UniGene ID:
Hs.591003
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
33110
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NIPA2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001008860.2
NP_001008860.1
6
920
NM_001008892.2
NP_001008892.1
5
784
NM_001008894.2
NP_001008894.1
4
727
NM_001184888.1
NP_001171817.1
5
875
NM_001184889.1
NP_001171818.1
9
1267
NM_030922.6
NP_112184.4
7
1042
XM_005272546.3
XP_005272603.1
7
1275
XM_005272547.4
XP_005272604.1
7
1059
XM_005272548.3
XP_005272605.1
6
889
XM_005272550.3
XP_005272607.1
6
727
XM_005272552.3
XP_005272609.1
5
989
XM_005272553.4
XP_005272610.1
5
793
XM_006720364.2
XP_006720427.1
8
1169
XM_006720365.2
XP_006720428.1
7
1014
XM_006720366.3
XP_006720429.1
6
880
XM_006720367.1
XP_006720430.1
6
1218
XM_011543877.2
XP_011542179.1
8
1402
XM_011543878.2
XP_011542180.1
8
1569
XM_011543879.2
XP_011542181.1
8
1140
XM_011543880.2
XP_011542182.1
6
1153
XM_017022645.1
XP_016878134.1
8
1463
XM_017022646.1
XP_016878135.1
8
1230
XM_017022647.1
XP_016878136.1
8
1178
XM_017022648.1
XP_016878137.1
8
1239
XM_017022649.1
XP_016878138.1
8
1373
XM_017022650.1
XP_016878139.1
8
1336
XM_017022651.1
XP_016878140.1
9
1757
XM_017022652.1
XP_016878141.1
8
1149
XM_017022653.1
XP_016878142.1
9
1463
XM_017022654.1
XP_016878143.1
7
987
XM_017022655.1
XP_016878144.1
6
985
XM_017022656.1
XP_016878145.1
6
995
XM_017022657.1
XP_016878146.1
7
1112
XM_017022658.1
XP_016878147.1
5
833
XM_017022659.1
XP_016878148.1
7
1083
XM_017022660.1
XP_016878149.1
5
1096
XM_017022661.1
XP_016878150.1
5
863
XM_017022662.1
XP_016878151.1
4
932
XM_017022663.1
XP_016878152.1
4
737
GenBank mRNA
AK096305.1
-
6
901
AK300843.1
-
4
670
AK313584.1
-
5
859
AY732242.1
-
9
1188
BC000957.3
-
2
213
BC011775.2
-
5
696
3640970
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
magnesium ion transport
magnesium ion transmembrane transport
magnesium ion transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0