Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278647.1 | NP_001265576.1 | 3 | 900 | ||
NM_001278648.1 | NP_001265577.1 | 3 | 921 | |||
NM_005341.3 | NP_005332.1 | 3 | 987 | |||
XM_017001110.1 | XP_016856599.1 | 3 | 972 | |||
XM_017001111.1 | XP_016856600.1 | 3 | 972 | |||
XM_017001112.1 | XP_016856601.1 | 3 | 977 | |||
XM_017001113.1 | XP_016856602.1 | 3 | 918 | |||
XM_017001114.1 | XP_016856603.1 | 3 | 905 | |||
GenBank mRNA | AK290156.1 | - | 3 | 895 | ||
AK312786.1 | - | 3 | 903 | |||
BC013573.2 | - | 2 | 846 | 3906038 | ||
L16896.1 | - | 3 | 927 |