Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031421.3 | NP_113609.1 | 2 | 238 | ||
NR_110662.1 | 2 | 238 | ||||
XM_011525323.2 | XP_011523625.1 | 2 | 256 | |||
XM_011525324.2 | XP_011523626.1 | 2 | 205 | |||
GenBank mRNA | AK055498.1 | - | 2 | 183 | ||
AL136760.1 | - | 2 | 205 | |||
BC067297.1 | - | 2 | 197 | 30383608 | ||
CF593846.1 | - | 2 | 254 |