Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257137.2 | NP_001244066.1 | 4 | 353 | ||
NM_001324197.1 | NP_001311126.1 | 4 | 374 | |||
NM_001324198.1 | NP_001311127.1 | 4 | 374 | |||
NM_031483.6 | NP_113671.3 | 4 | 353 | |||
XM_005260570.1 | XP_005260627.1 | 4 | 315 | |||
XM_011529079.1 | XP_011527381.1 | 2 | 139 | |||
XM_017028089.1 | XP_016883578.1 | 3 | 186 | |||
XM_017028090.1 | XP_016883579.1 | 5 | 429 | |||
XM_017028091.1 | XP_016883580.1 | 3 | 224 | |||
XM_017028092.1 | XP_016883581.1 | 5 | 429 | |||
GenBank mRNA | AB056663.2 | - | 4 | 263 | ||
AF095745.1 | - | 2 | 146 | |||
AK297809.1 | - | 4 | 315 | |||
AK315212.1 | - | 4 | 312 | |||
AL705688.1 | - | 4 | 354 | |||
BC011571.1 | - | 4 | 276 | 4560366 | ||
DC307047.1 | - | 4 | 332 |