Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAP2 SIRNA Assays ›
Gene Symbol:
MAP2
Gene Name:
microtubule associated protein 2
Gene Aliases:
MAP2A, MAP2B, MAP2C
Chromosome Location:
Chr.2: 209424047 - 209734118 on Build GRCh38
UniGene ID:
Hs.368281
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
33261
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039538.1
NP_001034627.1
5
615
NM_002374.3
NP_002365.3
4
412
NM_031845.2
NP_114033.2
4
412
NM_031847.2
NP_114035.2
4
412
XM_005246555.1
XP_005246612.1
4
531
XM_006712532.1
XP_006712595.1
4
531
XM_011511186.1
XP_011509488.1
4
438
XM_011511187.1
XP_011509489.1
3
335
XM_011511188.1
XP_011509490.1
3
335
XM_011511190.1
XP_011509492.1
3
335
XM_011511191.1
XP_011509493.1
3
335
XM_011511192.1
XP_011509494.1
3
335
XM_011511193.1
XP_011509495.1
3
335
XM_011511195.1
XP_011509497.1
3
335
XM_011511196.2
XP_011509498.1
4
531
XM_011511197.2
XP_011509499.1
4
531
XM_017004111.1
XP_016859600.1
3
335
XM_017004112.1
XP_016859601.1
4
531
XM_017004113.1
XP_016859602.1
4
438
XM_017004114.1
XP_016859603.1
4
531
XM_017004115.1
XP_016859604.1
3
335
XM_017004116.1
XP_016859605.1
3
335
XM_017004117.1
XP_016859606.1
3
335
XM_017004118.1
XP_016859607.1
4
438
XM_017004119.1
XP_016859608.1
4
531
XM_017004120.1
XP_016859609.1
4
531
XM_017004121.1
XP_016859610.1
4
438
XM_017004122.1
XP_016859611.1
4
438
XM_017004123.1
XP_016859612.1
4
531
XM_017004124.1
XP_016859613.1
4
438
XM_017004125.1
XP_016859614.1
4
531
XM_017004126.1
XP_016859615.1
4
531
XM_017004127.1
XP_016859616.1
4
531
XM_017004128.1
XP_016859617.1
3
335
XM_017004129.1
XP_016859618.1
3
335
XM_017004138.1
XP_016859627.1
4
438
XM_017004139.1
XP_016859628.1
1
195
XM_017004140.1
XP_016859629.1
4
438
GenBank mRNA
AB209330.1
-
3
330
AK291446.1
-
1
201
AK307603.1
-
4
390
BC027583.1
-
3
315
BC038857.1
-
5
604
BC066648.1
-
5
553
BC110423.1
-
4
660
BC117123.1
-
2
241
BC143245.1
-
2
246
U01828.1
-
4
412
U89329.1
-
1
164
U89330.1
-
1
164
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
microtubule bundle formation
central nervous system neuron development
neuron projection development
dendrite morphogenesis
dystroglycan binding
structural molecule activity
protein binding
calmodulin binding
microtubule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-77psw:80/100.66.77.247:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline