Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031907.1 | NP_114113.1 | 1 | 256 | ||
XM_017029892.1 | XP_016885381.1 | 2 | 342 | |||
GenBank mRNA | AF285593.1 | - | 1 | 256 | ||
AK292527.1 | - | 6 | 674 | |||
BC069073.1 | - | 1 | 255 | 7216887 | ||
BC101189.2 | - | 1 | 254 | 40019498 | ||
BC101190.2 | - | 1 | 254 | 40019499 | ||
BC101191.2 | - | 1 | 254 | 40019500 |