Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145646.1 | NP_001139118.1 | 5 | 779 | ||
NM_031301.3 | NP_112591.2 | 6 | 902 | |||
GenBank mRNA | AF508794.1 | - | 6 | 842 | ||
AK291204.1 | - | 6 | 844 | |||
AK307456.1 | - | 5 | 492 | |||
AK307725.1 | - | 6 | 964 | |||
AK310843.1 | - | 3 | 405 | |||
AL136671.1 | - | 6 | 848 | |||
AY358698.1 | - | 6 | 836 | |||
BC020905.1 | - | 6 | 844 | 4720647 |