Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005741.4 | NP_005732.2 | 6 | 3020 | ||
XM_006720831.1 | XP_006720894.1 | 6 | 3088 | |||
XM_006720832.1 | XP_006720895.1 | 5 | 3010 | |||
XM_006720833.1 | XP_006720896.1 | 4 | 2872 | |||
XM_006720834.3 | XP_006720897.1 | 5 | 3143 | |||
GenBank mRNA | BC008805.2 | - | 6 | 3018 | 3945548 | |
D88827.1 | - | 6 | 2980 |