Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017440.2 | NP_001017440.1 | 4 | 808 | ||
NM_031468.3 | NP_113656.2 | 5 | 798 | |||
XM_011516594.2 | XP_011514896.1 | 4 | 650 | |||
XM_011516595.1 | XP_011514897.1 | 4 | 398 | |||
XM_011516596.2 | XP_011514898.1 | 5 | 1293 | |||
XM_011516597.2 | XP_011514899.1 | 4 | 1226 | |||
XM_011516599.1 | XP_011514901.1 | 4 | 597 | |||
XM_017012675.1 | XP_016868164.1 | 6 | 1153 | |||
XM_017012676.1 | XP_016868165.1 | 5 | 935 | |||
XM_017012677.1 | XP_016868166.1 | 4 | 448 | |||
XM_017012678.1 | XP_016868167.1 | 5 | 910 | |||
XM_017012679.1 | XP_016868168.1 | 4 | 1263 | |||
XM_017012680.1 | XP_016868169.1 | 4 | 562 | |||
XM_017012681.1 | XP_016868170.1 | 4 | 425 | |||
XM_017012682.1 | XP_016868171.1 | 4 | 975 | |||
XM_017012683.1 | XP_016868172.1 | 5 | 759 | |||
GenBank mRNA | AF282250.1 | - | 5 | 529 | ||
AK123862.1 | - | 4 | 343 | |||
AK313054.1 | - | 4 | 398 | |||
AY007302.1 | - | 4 | 456 | |||
AY726595.1 | - | 3 | 284 | |||
BC020200.1 | - | 4 | 580 | 3944302 |