Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM107B SIRNA Assays ›
Gene Symbol:
FAM107B
Gene Name:
family with sequence similarity 107 member B
Gene Aliases:
C10orf45
Chromosome Location:
Chr.10: 14518557 - 14774897 on Build GRCh38
UniGene ID:
Hs.446315
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
33405
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM107B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282695.1
NP_001269624.1
6
3344
NM_001282696.1
NP_001269625.1
5
2820
NM_001282697.1
NP_001269626.1
4
2754
NM_001282698.1
NP_001269627.1
5
3068
NM_001282699.1
NP_001269628.1
4
3002
NM_001282700.1
NP_001269629.1
4
2720
NM_001282701.1
NP_001269630.1
4
2841
NM_001282702.1
NP_001269631.1
5
2907
NM_001282703.1
NP_001269632.1
4
2774
NM_001320735.1
NP_001307664.1
5
2786
NM_001320736.1
NP_001307665.1
5
3046
NM_001320737.1
NP_001307666.1
5
2840
NM_001320738.1
NP_001307667.1
5
2806
NM_001320739.1
NP_001307668.1
4
2806
NM_001320740.1
NP_001307669.1
4
2598
NM_001320741.1
NP_001307670.1
4
2699
NM_031453.3
NP_113641.2
5
3278
XM_005252616.3
XP_005252673.1
5
2675
XM_011519735.1
XP_011518037.1
6
2792
XM_017016746.1
XP_016872235.1
5
2666
XM_017016747.1
XP_016872236.1
4
2643
XM_017016748.1
XP_016872237.1
4
2678
XM_017016749.1
XP_016872238.1
4
2740
GenBank mRNA
AK024648.1
-
1
421
AK074275.1
-
3
2538
AK127413.1
-
5
3278
AL136885.1
-
4
2764
Back To Top
More Information
Gene Ontology Categories:
Function(s)
sensory perception of sound
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline