Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017440.2 | NP_001017440.1 | 6 | 1445 | ||
NM_031468.3 | NP_113656.2 | 7 | 1435 | |||
XM_011516594.2 | XP_011514896.1 | 6 | 1287 | |||
XM_011516595.1 | XP_011514897.1 | 6 | 1035 | |||
XM_011516596.2 | XP_011514898.1 | 7 | 1930 | |||
XM_011516597.2 | XP_011514899.1 | 6 | 1863 | |||
XM_011516599.1 | XP_011514901.1 | 6 | 1234 | |||
XM_017012675.1 | XP_016868164.1 | 8 | 1790 | |||
XM_017012676.1 | XP_016868165.1 | 7 | 1572 | |||
XM_017012677.1 | XP_016868166.1 | 6 | 1085 | |||
XM_017012678.1 | XP_016868167.1 | 7 | 1547 | |||
XM_017012679.1 | XP_016868168.1 | 6 | 1900 | |||
XM_017012680.1 | XP_016868169.1 | 6 | 1199 | |||
XM_017012681.1 | XP_016868170.1 | 6 | 1062 | |||
XM_017012682.1 | XP_016868171.1 | 6 | 1612 | |||
XM_017012683.1 | XP_016868172.1 | 7 | 1396 | |||
GenBank mRNA | AF282250.1 | - | 7 | 1166 | ||
AK123862.1 | - | 6 | 980 |