Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAP2 SIRNA Assays ›
Gene Symbol:
MAP2
Gene Name:
microtubule associated protein 2
Gene Aliases:
MAP2A, MAP2B, MAP2C
Chromosome Location:
Chr.2: 209424047 - 209734118 on Build GRCh38
UniGene ID:
Hs.368281
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
33433
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039538.1
NP_001034627.1
15
2554
NM_002374.3
NP_002365.3
15
6155
NM_031845.2
NP_114033.2
12
2087
NM_031847.2
NP_114035.2
13
2180
XM_005246555.1
XP_005246612.1
18
6784
XM_005246565.1
XP_005246622.1
14
6073
XM_005246566.1
XP_005246623.1
14
6061
XM_006712532.1
XP_006712595.1
18
6781
XM_011511186.1
XP_011509488.1
18
6691
XM_011511187.1
XP_011509489.1
17
6585
XM_011511188.1
XP_011509490.1
17
6585
XM_011511190.1
XP_011509492.1
16
6495
XM_011511191.1
XP_011509493.1
16
6453
XM_011511192.1
XP_011509494.1
16
6417
XM_011511193.1
XP_011509495.1
16
6414
XM_011511195.1
XP_011509497.1
16
6339
XM_011511196.2
XP_011509498.1
17
6526
XM_011511197.2
XP_011509499.1
17
6523
XM_017004111.1
XP_016859600.1
16
6414
XM_017004112.1
XP_016859601.1
17
6538
XM_017004113.1
XP_016859602.1
17
6430
XM_017004114.1
XP_016859603.1
16
6367
XM_017004115.1
XP_016859604.1
15
6159
XM_017004116.1
XP_016859605.1
15
6156
XM_017004117.1
XP_016859606.1
15
6156
XM_017004118.1
XP_016859607.1
15
6181
XM_017004119.1
XP_016859608.1
15
6271
XM_017004120.1
XP_016859609.1
15
6271
XM_017004121.1
XP_016859610.1
15
6178
XM_017004122.1
XP_016859611.1
15
6178
XM_017004123.1
XP_016859612.1
15
6262
XM_017004124.1
XP_016859613.1
15
6169
XM_017004125.1
XP_016859614.1
15
6259
XM_017004126.1
XP_016859615.1
15
6259
XM_017004127.1
XP_016859616.1
15
6259
XM_017004128.1
XP_016859617.1
14
6060
XM_017004129.1
XP_016859618.1
14
6057
XM_017004130.1
XP_016859619.1
14
6510
XM_017004131.1
XP_016859620.1
13
5902
XM_017004132.1
XP_016859621.1
13
5890
XM_017004133.1
XP_016859622.1
12
5809
XM_017004134.1
XP_016859623.1
12
5797
XM_017004135.1
XP_016859624.1
12
6243
XM_017004136.1
XP_016859625.1
12
5806
XM_017004137.1
XP_016859626.1
12
5794
XM_017004138.1
XP_016859627.1
14
2377
XM_017004139.1
XP_016859628.1
11
2134
XM_017004140.1
XP_016859629.1
12
2113
GenBank mRNA
AB209330.1
-
11
2005
AF088065.1
-
1
38
AL833567.1
-
6
1572
AV718243.1
-
1
342
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
microtubule bundle formation
central nervous system neuron development
neuron projection development
dendrite morphogenesis
dystroglycan binding
structural molecule activity
protein binding
calmodulin binding
microtubule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-9xzrh:80/100.66.79.6:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline