Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FGFBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031950.3 | NP_114156.1 | 1 | 183 | ||
GenBank mRNA | AB021123.1 | - | 1 | 154 | ||
AF361936.1 | - | 1 | 76 | |||
AY358978.1 | - | 1 | 183 | |||
BC025720.1 | - | 1 | 152 | 5228917 |