Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TATDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317889.1 | NP_001304818.1 | 3 | 149 | ||
NM_001317891.1 | NP_001304820.1 | 2 | 83 | |||
NM_032026.3 | NP_114415.1 | 3 | 149 | |||
NR_027427.1 | 3 | 149 | ||||
XM_006716666.3 | XP_006716729.1 | 3 | 169 | |||
XM_006716667.3 | XP_006716730.1 | 3 | 138 | |||
XM_006716668.3 | XP_006716731.1 | 3 | 151 | |||
XM_011517331.2 | XP_011515633.1 | 2 | 65 | |||
XM_017013895.1 | XP_016869384.1 | 3 | 159 | |||
XM_017013896.1 | XP_016869385.1 | 3 | 138 | |||
XM_017013897.1 | XP_016869386.1 | 2 | 59 | |||
XM_017013899.1 | XP_016869388.1 | 3 | 145 | |||
GenBank mRNA | AF212250.1 | - | 3 | 132 | ||
AK312204.1 | - | 3 | 145 | |||
BC064964.1 | - | 3 | 133 | 6141834 | ||
BG529611.1 | - | 2 | 74 | |||
BX332564.2 | - | 3 | 131 |