Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PPP1R12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002481.3 | NP_002472.2 | 14 | 2036 | ||
NM_032103.2 | NP_115286.1 | 2 | 391 | |||
NM_032104.2 | NP_115287.1 | 2 | 391 | |||
XM_017001342.1 | XP_016856831.1 | 15 | 2219 | |||
XM_017001343.1 | XP_016856832.1 | 15 | 2219 | |||
XM_017001344.1 | XP_016856833.1 | 15 | 2219 | |||
XM_017001345.1 | XP_016856834.1 | 15 | 2219 | |||
XM_017001346.1 | XP_016856835.1 | 14 | 2036 | |||
XM_017001347.1 | XP_016856836.1 | 14 | 2036 | |||
XM_017001348.1 | XP_016856837.1 | 14 | 2036 | |||
XM_017001349.1 | XP_016856838.1 | 15 | 2219 | |||
XM_017001350.1 | XP_016856839.1 | 15 | 2093 | |||
XM_017001352.1 | XP_016856841.1 | 5 | 486 | |||
XM_017001353.1 | XP_016856842.1 | 5 | 476 | |||
XM_017001354.1 | XP_016856843.1 | 2 | 1870 | |||
GenBank mRNA | AB003062.1 | - | 14 | 2049 | ||
AB050641.1 | - | 2 | 391 | |||
AB050642.1 | - | 2 | 391 | |||
AF324888.1 | - | 14 | 2030 | |||
AK307591.1 | - | 14 | 2036 | |||
BC144699.1 | - | 14 | 2028 | 9053233 | ||
BX501884.1 | - | 2 | 352 |