Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QTNF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031910.3 | NP_114116.3 | 3 | 694 | ||
NM_182486.1 | NP_872292.1 | 3 | 694 | |||
XM_011529857.1 | XP_011528159.1 | 4 | 3551 | |||
XM_017028569.1 | XP_016884058.1 | 3 | 2843 | |||
XM_017028570.1 | XP_016884059.1 | 5 | 1461 | |||
XM_017028571.1 | XP_016884060.1 | 5 | 3199 | |||
XM_017028572.1 | XP_016884061.1 | 5 | 3188 | |||
XM_017028573.1 | XP_016884062.1 | 5 | 977 | |||
XM_017028574.1 | XP_016884063.1 | 4 | 770 | |||
GenBank mRNA | AF329842.1 | - | 3 | 632 | ||
AK128125.1 | - | 8 | 4046 | |||
AK226149.1 | - | 2 | 1217 | |||
AK314060.1 | - | 3 | 651 | |||
AY358374.1 | - | 5 | 831 | |||
BC020551.1 | - | 3 | 693 | 2900683 | ||
BU535692.1 | - | 2 | 280 | |||
CR456451.1 | - | 3 | 644 |