Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETDB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160308.2 | NP_001153780.1 | 5 | 1145 | ||
NM_001320699.1 | NP_001307628.1 | 5 | 824 | |||
NM_031915.2 | NP_114121.2 | 6 | 1181 | |||
GenBank mRNA | AF334407.1 | - | 6 | 1100 | ||
AK307593.1 | - | 4 | 494 | |||
AK308220.1 | - | 5 | 1064 | |||
AK308354.1 | - | 4 | 642 | |||
BC028202.1 | - | 4 | 618 | |||
BC047434.2 | - | 4 | 581 | 5266911 |
Gene Symbol: SETDB2-PHF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320727.1 | NP_001307656.1 | 5 | 1145 | ||
NR_135324.1 | 5 | 1145 |