Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: HDAC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159286.1 | NP_001152758.1 | 13, 14 | 1743 | ||
NM_032019.5 | NP_114408.3 | 14, 15 | 1803 | |||
GenBank mRNA | AF393962.1 | - | 14, 15 | 1430 | ||
AF407272.1 | - | 14, 15 | 1430 | |||
AF407273.1 | - | 13, 14 | 1370 | |||
AF426160.1 | - | 14, 15 | 1783 | |||
AY450395.1 | - | 11, 12 | 1195 | |||
BC032049.1 | - | 14, 15 | 1616 | |||
BC094734.1 | - | 12, 13 | 1471 | |||
BC125082.1 | - | 8, 9 | 952 | |||
BC125083.1 | - | 14, 15 | 1477 | 40117772 | ||
CR456465.1 | - | 14, 15 | 1486 |