Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM234A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284497.1 | NP_001271426.1 | 3 | 548 | ||
NM_032039.3 | NP_114428.1 | 3 | 555 | |||
NR_104317.1 | 3 | 555 | ||||
XM_005255622.1 | XP_005255679.1 | 3 | 329 | |||
XM_005255623.2 | XP_005255680.1 | 3 | 277 | |||
XM_006720957.1 | XP_006721020.1 | 3 | 299 | |||
XM_011522691.1 | XP_011520993.1 | 4 | 322 | |||
XM_011522692.1 | XP_011520994.1 | 3 | 448 | |||
XM_017023760.1 | XP_016879249.1 | 3 | 665 | |||
XM_017023761.1 | XP_016879250.1 | 4 | 438 | |||
XM_017023762.1 | XP_016879251.1 | 4 | 424 | |||
XM_017023763.1 | XP_016879252.1 | 3 | 456 | |||
XM_017023764.1 | XP_016879253.1 | 3 | 329 | |||
XM_017023765.1 | XP_016879254.1 | 3 | 277 | |||
GenBank mRNA | AK057165.1 | - | 3 | 299 | ||
AK294581.1 | - | 3 | 267 | |||
AL136542.1 | - | 3 | 268 | |||
BC010521.1 | - | 1 | 26 | |||
BC013047.1 | - | 2 | 248 | 3857041 | ||
BC032112.2 | - | 2 | 248 | 4844625 | ||
CN262982.1 | - | 3 | 555 |