Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039091.2 | NP_001034180.1 | 1 | 238 | ||
NM_002765.4 | NP_002756.1 | 1 | 238 | |||
GenBank mRNA | AK313246.1 | - | 1 | 210 | ||
BC030019.2 | - | 1 | 179 | |||
BC040483.1 | - | 1 | 181 | 5272983 | ||
BC110875.1 | - | 1 | 172 | 6472836 | ||
BC119662.1 | - | 1 | 170 | 40116022 | ||
BC119663.1 | - | 1 | 170 | 40116023 | ||
CR988262.1 | - | 1 | 187 | |||
DA478574.1 | - | 1 | 238 | |||
Y00971.1 | - | 1 | 155 |