Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159286.1 | NP_001152758.1 | 15, 16 | 1895 | ||
NM_032019.5 | NP_114408.3 | 16, 17 | 1955 | |||
GenBank mRNA | AF393962.1 | - | 16, 17 | 1582 | ||
AF407272.1 | - | 16, 17 | 1582 | |||
AF407273.1 | - | 15, 16 | 1522 | |||
AF426160.1 | - | 16, 17 | 1935 | |||
AY450395.1 | - | 13, 14 | 1347 | |||
BC032049.1 | - | 16, 17 | 1768 | |||
BC094734.1 | - | 14, 15 | 1623 | |||
BC125082.1 | - | 10, 11 | 1104 | |||
BC125083.1 | - | 16, 17 | 1629 | 40117772 | ||
CR456465.1 | - | 16, 17 | 1638 |