Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032027.2 | NP_114416.1 | 7 | 1203 | ||
NR_135160.1 | 8 | 1333 | ||||
NR_135161.1 | 6 | 1129 | ||||
NR_135162.1 | 8 | 1330 | ||||
GenBank mRNA | AF353990.1 | - | 7 | 1201 | ||
BC029486.1 | - | 7 | 921 | 5271098 | ||
BC048995.1 | - | 7 | 919 |