Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077263.2 | NP_001070731.1 | 13 | 2460 | ||
NM_001206789.1 | NP_001193718.1 | 12 | 2355 | |||
GenBank mRNA | AB048797.1 | - | 12 | 2307 | ||
BC018715.1 | - | 14 | 2197 | |||
BC114928.1 | - | 13 | 2379 | 40028749 |