Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTNBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271667.1 | NP_001258596.1 | 7 | 734 | ||
NM_001271668.1 | NP_001258597.1 | 6 | 638 | |||
NM_001271669.1 | NP_001258598.1 | 5 | 584 | |||
NM_032122.4 | NP_115498.2 | 7 | 689 | |||
NM_183040.2 | NP_898861.1 | 7 | 689 | |||
NR_036448.1 | 8 | 828 | ||||
XM_005249447.4 | XP_005249504.1 | 8 | 690 | |||
XM_011514936.2 | XP_011513238.1 | 7 | 680 | |||
GenBank mRNA | AF061734.1 | - | 3 | 176 | ||
AF394226.1 | - | 7 | 606 | |||
AK054593.1 | - | 7 | 658 | |||
AK290718.1 | - | 7 | 606 | |||
AK310590.1 | - | 8 | 690 | |||
AL136637.2 | - | 7 | 606 | |||
AY265460.1 | - | 7 | 613 | |||
BC011912.2 | - | 7 | 604 | 4139934 | ||
BE793298.1 | - | 546 | ||||
BQ056746.1 | - | 6 | 577 | |||
BX394617.2 | - | 8 | 828 | |||
KU178801.1 | - | 7 | 500 | |||
KU178802.1 | - | 5 | 395 |