Search Thermo Fisher Scientific
Gene Symbol: TXNDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098529.1 | NP_001091999.1 | 2 | 1297 | ||
NM_032243.5 | NP_115619.4 | 2 | 1094 | |||
XM_017026041.1 | XP_016881530.1 | 3 | 968 | |||
GenBank mRNA | AF080095.1 | - | 2 | 827 | ||
AK097656.1 | - | 2 | 1047 | |||
BC050132.1 | - | 2 | 882 | 5164861 | ||
EF560747.1 | - | 2 | 973 |