Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032152.4 | NP_115528.4 | 6 | 1792 | ||
XM_005272502.2 | XP_005272559.1 | 6 | 1778 | |||
XM_011528352.2 | XP_011526654.1 | 6 | 1798 | |||
XM_011528353.2 | XP_011526655.1 | 6 | 1796 | |||
GenBank mRNA | AJ272324.1 | - | 5 | 1936 | ||
AK308227.1 | - | 6 | 1775 | |||
BC028012.1 | - | 7 | 1749 | 5215850 |