Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006361.5 | NP_006352.2 | 2 | 893 | ||
GenBank mRNA | AK225234.1 | - | 2 | 837 | ||
AK313266.1 | - | 2 | 836 | |||
AY937237.1 | - | 2 | 902 | |||
BC007092.1 | - | 2 | 823 | 4250486 | ||
BC070233.1 | - | 2 | 828 | 6668163 | ||
BT007410.1 | - | 2 | 737 | |||
KF613154.1 | - | 2 | 737 | |||
U57052.1 | - | 2 | 791 | |||
U81599.1 | - | 2 | 913 |