Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286211.1 | NP_001273140.1 | 2 | 553 | ||
NM_001286212.1 | NP_001273141.1 | 2 | 553 | |||
NM_001286213.1 | NP_001273142.1 | 2 | 553 | |||
NM_032256.2 | NP_115632.1 | 2 | 553 | |||
XM_011538831.2 | XP_011537133.1 | 2 | 562 | |||
XM_011538832.2 | XP_011537134.1 | 2 | 237 | |||
XM_017020023.1 | XP_016875512.1 | 2 | 279 | |||
GenBank mRNA | AK057287.1 | - | 2 | 272 | ||
AL136855.1 | - | 2 | 255 | |||
BC030284.1 | - | 2 | 261 | 5240522 | ||
BC060798.1 | - | 2 | 316 | 30348706 | ||
CF994089.1 | - | 642 |