Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FYTTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011537.2 | NP_001011537.2 | 3 | 301 | ||
NM_032288.6 | NP_115664.2 | 2 | 360 | |||
NR_027840.1 | 2 | 471 | ||||
GenBank mRNA | AJ344094.1 | - | 2 | 206 | ||
AK027672.1 | - | 2 | 244 | |||
AK296283.1 | - | 2 | 175 | |||
AK302546.1 | - | 2 | 471 | |||
AK303462.1 | - | 3 | 301 | |||
AK315019.1 | - | 2 | 209 | |||
AK316308.1 | - | 2 | 101 | |||
AL136558.1 | - | 2 | 213 | |||
BC035006.2 | - | 2 | 363 | 5267824 | ||
BC039734.1 | - | 2 | 270 | 6042926 | ||
BX509179.1 | - | 2 | 210 | |||
CR533502.1 | - | 2 | 141 |