Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261467.1 | NP_001248396.1 | 3 | 209 | ||
NM_032141.3 | NP_115517.1 | 4 | 303 | |||
XM_011525345.2 | XP_011523647.1 | 4 | 218 | |||
XM_017025209.1 | XP_016880698.1 | 5 | 422 | |||
XM_017025210.1 | XP_016880699.1 | 4 | 1133 | |||
XM_017025211.1 | XP_016880700.1 | 5 | 331 | |||
XM_017025212.1 | XP_016880701.1 | 4 | 468 | |||
XM_017025213.1 | XP_016880702.1 | 3 | 248 | |||
XM_017025214.1 | XP_016880703.1 | 3 | 277 | |||
XM_017025215.1 | XP_016880704.1 | 3 | 281 | |||
GenBank mRNA | AK094552.1 | - | 1647 | |||
AK297102.1 | - | 3 | 204 | |||
AK313963.1 | - | 4 | 290 | |||
AL136806.1 | - | 4 | 294 | |||
BC016313.1 | - | 5 | 317 | |||
BC040118.1 | - | 4 | 283 | 5176197 | ||
BC062425.1 | - | 4 | 296 | |||
BC105044.1 | - | 4 | 290 | 8144047 | ||
BC105046.1 | - | 4 | 290 | 8144049 | ||
BC143539.1 | - | 3 | 196 | |||
CR533555.1 | - | 4 | 266 | |||
DC327004.1 | - | 3 | 209 |