Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032314.3 | NP_115690.3 | 2 | 238 | ||
XM_006719639.2 | XP_006719702.1 | 3 | 338 | |||
GenBank mRNA | AK057777.1 | - | 2 | 228 | ||
AK222610.1 | - | 2 | 228 | |||
AK225242.1 | - | 2 | 228 | |||
AK293656.1 | - | 2 | 259 | |||
AK298214.1 | - | 2 | 238 | |||
AK315681.1 | - | 2 | 229 | |||
BC004916.2 | - | 2 | 214 | |||
BC107874.1 | - | 2 | 232 | 6452052 | ||
BP307378.1 | - | 2 | 238 | |||
BX647562.1 | - | 2 | 227 | |||
EU700459.1 | - | 2 | 232 |