Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF414 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146175.1 | NP_001139647.1 | 2, 3 | 442 | ||
NM_032370.2 | NP_115746.2 | 2, 3 | 442 | |||
GenBank mRNA | AK074191.1 | - | 2, 3 | 438 | ||
AK225034.1 | - | 2, 3 | 438 | |||
BC007310.1 | - | 2, 3 | 426 | 3353224 | ||
BC071937.1 | - | 2, 3 | 421 | 5089259 |